Novus Biologicals
Manufacturer Code:NBP144886
Catalog # NBP144886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Bacterially expressed full-length human CtBP1 (NM_001012614.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BARS brefeldin A-ribosylated substrate CTBP CtBP1 C-terminal binding protein 1 C-terminal-binding protein 1 EC 1.1.1 EC 1.1.1.- EC 1.1.1.158 MGC104684; brefeldin A-ribosylated substrate; C-terminal-binding protein 1; CtBP1
Gene Aliases: BARS; CTBP; CTBP1
UniProt ID: (Human) Q13363
Entrez Gene ID: (Human) 1487
Molecular Function: dehydrogenase oxidoreductase transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.