R&D Systems
Manufacturer Code:AF2299
Catalog # AF2299
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Crossveinless-2/CV-2 (R&D Systems Catalog # 2299-CV) Ala39-Arg685 Accession # AAH66153 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP binding endothelial regulator BMP-binding endothelial regulator precursor protein BMP-binding endothelial regulator protein BMPER Bone morphogenetic protein-binding endothelial cell precursor-derived regulator CRIM3 crossveinless 2 crossveinless-2 Cv2 CV-2 hCV2 KIAA1965 Protein crossveinless-2; BMP-binding endothelial regulator precursor protein; BMP-binding endothelial regulator protein; Bone morphogenetic protein-binding endothelial cell precursor-derived regulator; crossveinless 2; crossveinless-2; hCV2; Protein crossveinless-2
Gene Aliases: BMPER; CRIM3; CV-2; CV2; KIAA1965
UniProt ID: (Human) Q8N8U9
Entrez Gene ID: (Human) 168667
Molecular Function: cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.