Invitrogen
Manufacturer Code:PA579067
Catalog # PIPA579067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human Crk p38 recombit protein (Position: A2-R246). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adapter molecule crk; Adapter molecule crk Proto-oncogene c-Crk p38 CRK; avian sarcoma virus CT10 (v-crk) oncogene homolog; CT-10 related kinase 3; p38; Proto-oncogene c-Crk; v-crk avian sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene-like protein
Gene Aliases: c-Crk; CRK; Crk-I; Crk-II; Crk-III; Crk3; CRKII; CrkIII; Crko; p38
UniProt ID: (Human) P46108, (Mouse) Q64010, (Rat) Q63768
Entrez Gene ID: (Human) 1398, (Mouse) 12928, (Rat) 54245
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.