Invitrogen
Manufacturer Code:PA534476
Catalog # PIPA534476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide near the amino terminus of human TDGF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CRGF; cripto growth factor; Cripto-1 growth factor; epidermal growth factor-like Cripto protein; Epidermal growth factor-like cripto protein CR1; TDGF1 Teratocarcinomaderived growth factor 1 Epidermal growth factor-like cripto protein CR1 Cripto-1 growth factor CRGF CR CRGF CRIPTO Cripto-1; teratocarcinoma-derived growth factor; Teratocarcinoma-derived growth factor 1; TERATOCARCINOMA-DERIVED GROWTH FACTOR PRECURSOR (EPIDERMAL GROWTH FACTOR-LIKE CRIPTO PROTEIN) (CRIPTO GROWTH FACTOR)
Gene Aliases: CR; CR1; CRGF; CRIPTO; TDGF1
UniProt ID: (Human) P13385
Entrez Gene ID: (Human) 6997, (Rat) 680246, (Mouse) 21667
Molecular Function: calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.