Invitrogen
Manufacturer Code:PA5113731
Catalog # PIPA5113731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CKM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-CK; BCK B-CK Brain creatine kinase CK MB Ck3 Ck-3 Ckb Ckbb Ckbr Ckm CKMM CPK-B CPK-M creatine kinase B Creatine kinase B chain creatine kinase brain creatine kinase brain-type Creatine kinase B-type Creatine kinase M chain Creatine kinase M-type creatine kinase brain creatine kinase M-type creatine kinase muscle creatine kinase-B creatine kinase-M Creatine phosphokinase B-type Creatine phosphokinase M-type epididymis luminal protein 211 epididymis secretory protein Li 29 HEL-211 HEL-S-29 MCK M-CK; Brain creatine kinase; CPK-B; CPK-M; Creatine kinase B chain; Creatine kinase B-type; creatine kinase brain; creatine kinase brain-type; Creatine kinase M chain; Creatine kinase M-type; creatine kinase, brain; creatine kinase, muscle; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; M-CK
Gene Aliases: B-CK; BCK; Ck-3; Ck3; CKB; CKBB; CKM; CKMM; HEL-211; HEL-S-29; M-CK; MCK
UniProt ID: (Human) P12277, (Human) P06732, (Mouse) Q04447, (Mouse) P07310
Entrez Gene ID: (Human) 1152, (Human) 1158, (Mouse) 12709, (Mouse) 12715
Molecular Function: amino acid kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.