R&D Systems
Manufacturer Code:MAB9076SP
Catalog # MAB9076SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 969409 |
Immunogen | <em>E. coli</em>-derived recombinant human Creatine Kinase BB Pro2-Lys381 Accession # P12277 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B-CK; B-CK CKB Creatine Kinase Isoenzyme; Brain creatine kinase; CPK-B; Creatine kinase B chain; Creatine kinase B-type; creatine kinase brain; creatine kinase brain-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; epididymis luminal protein 211; epididymis secretory protein Li 29
Gene Aliases: B-CK; BCK; CKB; CKBB; HEL-211; HEL-S-29
UniProt ID: (Human) P12277
Entrez Gene ID: (Human) 1152
Molecular Function: amino acid kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.