Novus Biologicals
Manufacturer Code:NBP237621
Catalog # NBP237621
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4C6 |
Immunogen | Purified recombinant fragment of human CTTN expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1110020L01Rik; 1110020L01Rik Amplaxin cortactin ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 srcsubstrate) EMS1amplaxin FLJ34459 Oncogene EMS1 src substrate cortactin; Amplaxin; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); Oncogene EMS1; Src substrate cortactin
Gene Aliases: CTTN; EMS1
UniProt ID: (Human) Q14247
Entrez Gene ID: (Human) 2017
Molecular Function: actin family cytoskeletal protein basic helix-loop-helix transcription factor cytoskeletal protein non-motor actin binding protein transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.