Invitrogen
Manufacturer Code:PA529799
Catalog # PIPA529799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 276 of Human Cortactin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110020L01Rik; Amplaxin; EMS1 1110020L01Rik amplaxin ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate) oncogene EMS1 src substrate cortactin; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); Oncogene EMS1; Src substrate cortactin
Gene Aliases: CTTN; EMS1
UniProt ID: (Human) Q14247
Entrez Gene ID: (Human) 2017
Molecular Function: actin family cytoskeletal protein basic helix-loop-helix transcription factor cytoskeletal protein non-motor actin binding protein transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.