Proteintech
Manufacturer Code:113811AP150UL
Catalog # 50-172-7110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cortactin Fusion Protein Ag1940 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110020L01Rik; Amplaxin; Amplaxin cortactin CTTN EMS1 Oncogene EMS1 Src substrate cortactin; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); mammary tumor and squamous cell carcinoma associated (p80/85 src substrate); Oncogene EMS1; Src substrate cortactin
Gene Aliases: 1110020L01Rik; CTTN; Cttnb; EMS1
UniProt ID: (Human) Q14247, (Mouse) Q60598, (Rat) Q66HL2
Entrez Gene ID: (Human) 2017, (Mouse) 13043, (Rat) 60465
Molecular Function:
actin family cytoskeletal protein
basic helix-loop-helix transcription factor
cytoskeletal protein
non-motor actin binding protein
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.