Invitrogen
Manufacturer Code:PA521799
Catalog # PIPA521799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 213 of Coronin 1B (Uniprot IDNo.Q9BR76) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coronin 1b; coronin 2; coronin, actin binding protein, 1B; Coronin-1B; Coronin-2; CORONIN-2 coronin-1B
Gene Aliases: CORO1B; CORONIN-2
UniProt ID: (Human) Q9BR76, (Mouse) Q9WUM3
Entrez Gene ID: (Human) 57175, (Mouse) 23789
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.