Invitrogen
Manufacturer Code:PA529875
Catalog # PIPA529875
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 125 and 364 of Human Coronin 1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLABP CLIPINA HCORO1 TACO p57 clipin-A coronin-1 coronin-1A coronin-like protein A coronin-like protein p57 tryptophan aspartate-containing coat protein; Clipin-A; coronin, actin binding protein, 1A; coronin-1; Coronin-1A; Coronin-like protein A; Coronin-like protein p57; TACO; Tryptophan aspartate-containing coat protein
Gene Aliases: CLABP; CLIPINA; CORO1; CORO1A; HCORO1; IMD8; p57; TACO
UniProt ID: (Human) P31146
Entrez Gene ID: (Human) 11151
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.