Novus Biologicals
Manufacturer Code:NBP179763
Catalog # NBP179763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The specific Immunogen is proprietary information. Peptide sequence YLQALRTVGGICQDYDSDKRFPAFGFGARIPPNFEVSHDFAINFDPENPE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Copine VI; copine VI (neuronal); Copine VI copine VI (neuronal) copine-6 N-COPINE neuronal copine Neuronal-copine; Copine-6; N-copine; Neuronal-copine
Gene Aliases: CPNE6
UniProt ID: (Human) O95741
Entrez Gene ID: (Human) 9362
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.