Invitrogen
Manufacturer Code:701297
Catalog # PI701297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 20H24L10 |
Immunogen | Peptide corresponding to amino acids 126164 of human copeptin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: ADH ARVP AVP-NPII AVRP VP antidiuretic hormone neurohypophyseal vasopressin-neurophysin 2-copeptin vasopressin-neurophysin II-copeptin; antidiuretic hormone; Arg-vasopressin; Arginine-vasopressin; AVP-NPII; Copeptin; neurohypophyseal; Neurophysin 2; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; Vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin
Gene Aliases: ADH; ARVP; AVP; AVP-NPII; AVRP; VP
UniProt ID: (Human) P01185
Entrez Gene ID: (Human) 551
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.