R&D Systems
Manufacturer Code:MAB7737SP
Catalog # MAB7737SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 578618 |
Immunogen | Mouse myeloma cell line NS0 transfected with human Connexin 43/GJA1 Accession # P17302 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: connexin 43; connexin 43 connexin-43 Cx43 CX43gap junction protein alpha-like DFNB38 Gap junction 43 kDa heart protein gap junction alpha-1 protein gap junction protein alpha 1 43kDa gap junction protein alpha 1 43kDa (connexin 43) GJA1 GJAL HSS ODD ODDD ODOD SDTY3; Connexin-43; Cx43; Gap junction 43 kDa heart protein; Gap junction alpha-1 protein; gap junction protein, alpha 1, 43kDa
Gene Aliases: AVSD3; CMDR; CX43; EKVP; GJA1; GJAL; HLHS1; HSS; ODDD; PPKCA
UniProt ID: (Human) P17302
Entrez Gene ID: (Human) 2697
Molecular Function:
cell junction protein
gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.