Novus Biologicals
Manufacturer Code:NBP289209JF549
Catalog # NB087741
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 105 |
Immunogen | This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Connexin 43/GJA1. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: connexin 43; connexin 43 connexin-43 Cx43 CX43gap junction protein alpha-like DFNB38 Gap junction 43 kDa heart protein gap junction alpha-1 protein gap junction protein alpha 1 43kDa gap junction protein alpha 1 43kDa (connexin 43) GJAL HSS ODD ODDD ODOD SDTY3; Connexin-43; Cx43; Gap junction 43 kDa heart protein; Gap junction alpha-1 protein; gap junction protein, alpha 1, 43kDa
Gene Aliases: AVSD3; CMDR; CX43; EKVP; GJA1; GJAL; HLHS1; HSS; ODDD; PPKCA
UniProt ID: (Human) P17302
Entrez Gene ID: (Human) 2697
Molecular Function:
cell junction protein
gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.