Novus Biologicals
Manufacturer Code:NB10081867
Catalog # NB10081867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around aa. 365~369 (R-A-S-S-R) derived from Human Connexin 43. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: connexin 43; connexin 43 connexin-43 Cx43 CX43gap junction protein alpha-like DFNB38 Gap junction 43 kDa heart protein gap junction alpha-1 protein gap junction protein alpha 1 43kDa gap junction protein alpha 1 43kDa (connexin 43) GJAL HSS ODD ODDD ODOD SDTY3; Connexin-43; Cx43; Gap junction 43 kDa heart protein; Gap junction alpha-1 protein; gap junction protein, alpha 1, 43kDa
Gene Aliases: AVSD3; CMDR; CX43; EKVP; GJA1; GJAL; HLHS1; HSS; ODDD; PPKCA
UniProt ID: (Human) P17302
Entrez Gene ID: (Human) 2697
Molecular Function:
cell junction protein
gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.