Invitrogen
Manufacturer Code:PA125098
Catalog # PIPA125098
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH corresponding to residues K(362) P S S R A S S R A S S R P R P D D L E I(382) of human GJA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 1 connexin; connexin 43; Connexin-43; Cx43; CX43 DFNB38 GJAL HSS ODDD; Gap junction 43 kDa heart protein; Gap junction alpha-1 protein; gap junction membrane channel protein alpha 1; gap junction protein connexin 43; gap junction protein, alpha 1, 43 kD (connexin 43); gap junction protein, alpha 1, 43kDa; Vascular smooth muscle connexin-43; vascular smooth muscle connexin43
Gene Aliases: AU042049; AVSD3; AW546267; CMDR; Cnx43; connexin43; CX43; Cx43alpha1; Cxn-43; EKVP; Gja-1; GJA1; GJAL; HLHS1; HSS; Npm1; ODDD; PPKCA
UniProt ID: (Human) P17302, (Bovine) P18246, (Mouse) P23242, (Rat) P08050
Entrez Gene ID: (Human) 2697, (Bovine) 281193, (Mouse) 14609, (Rat) 24392, (Chinese hamster) 100689077
Molecular Function:
cell junction protein
gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.