Invitrogen
Manufacturer Code:PA137496
Catalog # 01-673-317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from a segment of the 3 cytoplasmic domain of rat connexin 43 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze,do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: connexin 43; Connexin-43; Cx43; Gap junction 43 kDa heart protein; Gap junction alpha-1 protein; gap junction membrane channel protein alpha 1; gap junction protein, alpha 1, 43kDa; GJA1 ODDD Oculodentodigital dysplasia CX43 DFNB38 Gap junction 43kDa heart protein SDTY3 Syndactyly type III
Gene Aliases: AVSD3; CMDR; CX43; Cxn-43; EKVP; GJA1; GJAL; HLHS1; HSS; ODDD; PPKCA
UniProt ID: (Human) P17302, (Rat) P08050
Entrez Gene ID: (Human) 2697, (Rat) 24392
Molecular Function: cell junction protein gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.