Novus Biologicals
Manufacturer Code:NBP30359020UL
Catalog # NB095542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 229-358 of human Connexin 40/GJA5 (NP_859054.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 5 40kD (connexin 40) connexin-40 Cx40 gap junction alpha-5 protein gap junction protein alpha 5 40kDa gap junction protein alpha 5 40kDa (connexin 40) MGC11185; connexin 40; Connexin-40; Cx40; Gap junction alpha-5 protein; gap junction protein alpha 5 40kDa
Gene Aliases: ATFB11; CX40; GJA5
UniProt ID: (Human) P36382
Entrez Gene ID: (Human) 2702
Molecular Function: cell junction protein gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.