Invitrogen
Manufacturer Code:PA577356
Catalog # PIPA577356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)GHRFPQGYHSDKR corresponding to amino acid residues 328-340 of rat Connexin-40 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: connexin 40; Connexin-40; CX40; Gap junction alpha-5 protein; gap junction membrane channel protein alpha 5
Gene Aliases: 5730555N10Rik; Cnx40; Cx40; Cxn-40; Gja-5; Gja5
UniProt ID: (Rat) P28234, (Mouse) Q01231
Entrez Gene ID: (Rat) 50563, (Mouse) 14613
Molecular Function:
cell junction protein
gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.