Invitrogen
Manufacturer Code:PA118406
Catalog # 01-670-656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues S(253) L V Q G L T P P P D F N Q C L K(269) of rat Connexin-40. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: connexin 40; Connexin-40; Cx40; Gap junction alpha-5 protein; Gap junction alpha-5 protein Cx40 Gja5 Cxn-40; gap junction membrane channel protein alpha 5
Gene Aliases: Cx40; Cxn-40; Gja5
UniProt ID: (Rat) P28234
Entrez Gene ID: (Rat) 50563
Molecular Function: cell junction protein gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.