Novus Biologicals
Manufacturer Code:NBP303264100UL
Catalog # NB093943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-134 of human Complexin-1 (NP_006642.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: complexin 1 Complexin I complexin-1 CPX I CPX1 CPX-I synaphin 2 Synaphin-2; Complexin I; Complexin-1; CPX I; synaphin 2; Synaphin-2
Gene Aliases: CPLX1; CPX-I; CPX1
UniProt ID: (Human) O14810
Entrez Gene ID: (Human) 10815
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.