Invitrogen
Manufacturer Code:PA589110
Catalog # PIPA589110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 80-420 of human CFB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative-complement pathway C3/C5 convertase; B-factor, properdin; C3/C5 convertase; complement component 2 (within H-2S); complement factor B; Factor B; histocompatibility 2, complement component factor B; properdin; properdin factor B
Gene Aliases: AI195813; AI255840; B; Bf; C2; Da1-24; Fb; H2-Bf
Entrez Gene ID: (Mouse) 14962, (Rat) 294257
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.