Invitrogen
Manufacturer Code:PA514244
Catalog # PIPA514244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 395-422 amino acids from the central region of human BF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative-complement pathway C3/C5 convertase; B-factor, properdin; C3 proaccelerator; C3 proactivator; C3/C5 convertase; complement component 2 (within H-2S); Complement factor B; Complement factor B Ba fragment; Complement factor B Bb fragment; Complement factor B C3/C5 convertase Properdin factor B Glycine-rich beta glycoprotein GBG PBF2 Complement factor B Ba fragment Complement factor B Bb fragment BFD BF CFB; Factor B; GBG; Glycine-rich beta glycoprotein; glycine-rich beta-glycoprotein; histocompatibility 2, complement component factor B; PBF2; properdin; Properdin factor B
Gene Aliases: AHUS4; AI195813; AI255840; ARMD14; B; BF; BFD; C2; CFAB; CFB; CFBD; FB; FBI12; GBG; H2-Bf; PBF2
UniProt ID: (Human) P00751
Entrez Gene ID: (Human) 629, (Mouse) 14962
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.