Novus Biologicals
Manufacturer Code:NBP189985
Catalog # NBP189985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KDNEQHVFKVKDMENLEDVFYQMIDESQSLSLCGMVWEHRKGTDYHKQPWQAKISVIRPSKGHESCMGAVVSEYFVLTAAHCFTVDDKEHSIKVSVGGEKRDLEIEVVLFHPNYNINGKKEAGIPEFYDYDVALIKLKNKLKYG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-factor, properdin; BF AHUS4 B-factor properdin BFD C3 proaccelerator C3 proactivator C3/C5 convertase complement factor B EC 3.4.21 EC 3.4.21.47 FB FBI12 GBGCFAB Glycine-rich beta glycoprotein glycine-rich beta-glycoprotein H2-Bf PBF2 Properdin factor B; C3 proaccelerator; C3 proactivator; C3/C5 convertase; Complement factor B; Complement factor B Ba fragment; Complement factor B Bb fragment; GBG; Glycine-rich beta glycoprotein; glycine-rich beta-glycoprotein; PBF2; Properdin factor B
Gene Aliases: AHUS4; ARMD14; BF; BFD; CFAB; CFB; CFBD; FB; FBI12; GBG; H2-Bf; PBF2
UniProt ID: (Human) P00751
Entrez Gene ID: (Human) 629
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.