Novus Biologicals
Manufacturer Code:NBP281062PE
Catalog # NB015178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G7 |
Immunogen | The 2e loop of the Complement Component C3aR. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AZ3B AZ3BC3a anaphylatoxin chemotactic receptor C3a-R C3AR C3AR1 C3ARcomplement component 3 receptor 1 C3R1 complement component 3a receptor 1 Complement Component C3a R Complement Component C3aR HNFAG09; C3a anaphylatoxin chemotactic receptor; C3AR; complement component 3 receptor 1
Gene Aliases: AZ3B; C3AR; C3AR1; C3R1; HNFAG09
UniProt ID: (Human) Q16581
Entrez Gene ID: (Human) 719
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.