Novus Biologicals
Manufacturer Code:NBP223498
Catalog # NBP223498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | C17/5 |
Immunogen | Purified human C5a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: anaphylatoxin C5a analog; anaphylatoxin C5a analog C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4 C5 complement C5 complement component 5 CPAMD4MGC142298 FLJ17816 FLJ17822; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C5a anaphylatoxin; Complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; prepro-C5
Gene Aliases: C5; C5a; C5b; C5D; CPAMD4; ECLZB
UniProt ID: (Human) P01031
Entrez Gene ID: (Human) 727
Molecular Function: complement component cytokine defense/immunity protein enzyme modulator protease inhibitor serine protease inhibitor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.