Novus Biologicals
Manufacturer Code:NBP2345810.1 MG
Catalog # NBP23458101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | C4D204 |
Immunogen | Recombinant human Complement 4d protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acidic C4; Acidic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C4A anaphylatoxin; C4b-A; C4d-A; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; MHC class III region complement; Rodgers form of C4
Gene Aliases: C4; C4A; C4A2; C4A3; C4A4; C4A6; C4AD; C4S; CO4; CPAMD2; RG
UniProt ID: (Human) P0C0L4
Entrez Gene ID: (Human) 720
Molecular Function: complement component cytokine defense/immunity protein enzyme modulator protease inhibitor serine protease inhibitor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.