Novus Biologicals
Manufacturer Code:NB200541
Catalog # NB200541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 16D2 |
Immunogen | Cell-bound C4 of mouse origin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Acidic complement C4; basic C4 Basic complement C4 C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3 C4B1 C4B12 C4B2 C4B3 C4FMGC164979 CH Chido form of C4 CO4C4B5 complement C4-B complement C4B1a complement component 4B complement component 4B (Chido blood group) CPAMD3FLJ60561 EC 2.1.1.144 EC 2.7.11; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; C4a anaphylatoxin; C4b-A; C4b-B; C4d-A; C4d-B; Chido form of C4; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a
Gene Aliases: C4A; C4B; C4B1; C4B12; C4B2; C4B3; C4B5; C4B_2; C4BD; C4F; CH; CO4; CPAMD2; CPAMD3
UniProt ID: (Human) P0C0L4
Entrez Gene ID: (Human) 721
Molecular Function:
complement component
cytokine
defense/immunity protein
enzyme modulator
protease inhibitor
serine protease inhibitor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.