Invitrogen
Manufacturer Code:LFMA0198
Catalog # PILFMA0198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 52H10 |
Immunogen | Protein purified from Human plasma |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; C4a anaphylatoxin; C4b-A; C4b-B; C4d-A; C4d-B; Chido form of C4; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; DADB-112B14.11 C4B1 C4B12 C4B2 C4B3 C4B5 C4F CH CO4 CPAMD3 FLJ60561 MGC164979
Gene Aliases: C4A; C4B; C4B1; C4B12; C4B2; C4B3; C4B5; C4B_2; C4BD; C4F; CH; CO4; CPAMD2; CPAMD3
UniProt ID: (Human) P0C0L4
Entrez Gene ID: (Human) 721
Molecular Function:
complement component
cytokine
defense/immunity protein
enzyme modulator
protease inhibitor
serine protease inhibitor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.