Invitrogen
Manufacturer Code:PA19567
Catalog # 01-674-443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoblot (IB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse C3 complement protein aa 678-692 part of C3a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylation stimulating protein; ASP; C3 Acylation Stimulating Protein Cleavage Product; C3-beta-c; C3a anaphylatoxin; C3adesArg; C3bc; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; HSE-MSF
Gene Aliases: AI255234; ASP; C3; HSE-MSF; Plp
UniProt ID: (Mouse) P01027
Entrez Gene ID: (Mouse) 12266
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.