Invitrogen
Manufacturer Code:MA535077
Catalog # PIMA535077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0541 |
Immunogen | A synthesized peptide derived from human C3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Acylation stimulating protein; Acylation stimulating protein AHUS5 AI255234 ARMD9 ASP C3 C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1 C3a C3a anaphylatoxin C3adesArg C3b C3bc C3-beta-c C3d Complement C3 Complement C3 alpha chain Complement C3 beta chain Complement C3b alpha&prime chain Complement C3c alpha&prime chain fragment 1 Complement C3c alpha&prime chain fragment 2 Complement C3d fragment Complement C3dg fragment Complement C3f fragment Complement C3g fragment CPAMD1 ENCF-1 ENCF-2 HEL-S-62p HSE-MSF I79_008227 Neutrophil chemotactic factor-1 Neutrophil chemotactic factor-2 Plp; acylation-stimulating protein cleavage product; ASP; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3-beta-c; C3a anaphylatoxin; C3adesArg; C3bc; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component C3a; complement component C3b; epididymis secretory sperm binding protein Li 62p; prepro-C3
Gene Aliases: AHUS5; ARMD9; ASP; C3; C3a; C3b; CPAMD1; HEL-S-62p
UniProt ID: (Human) P01024
Entrez Gene ID: (Human) 718
Molecular Function: complement component cytokine defense/immunity protein enzyme modulator protease inhibitor serine protease inhibitor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.