Test
Invitrogen
Catalog # HYB 005-01-02
Quantity:
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
Export as a spreadsheet
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer. |
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com.
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase.
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | 1:30,000 |
Western Blot (WB) | Assay-Dependent |
Host / Isotype | Mouse / IgG1, kappa |
Class | Monoclonal |
Type | Antibody |
Clone | 2D8 |
Immunogen | C3 isolated from human plasma. |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | 0.92 mg/mL |
Storage conditions | 4° C, store in dark |
RRID | AB_1073811 |
HYB 005-01-02 is specific for human C3.
HYB 005-01-02 has been successfully used in ELISA, and Western blotting procedures. HYB 005-01 reacts strongly with C3. Strong reaction is seen in ELISA with C3 directly coated onto the microtiter well, and also when tested in sandwich ELISA in combination with a polyclonal antibody against C3 (e.g. DAKO A 0062). In Western blotting after SDS-PAGE, HYB 005-01 reacts with C3 in reduced as well as non-reduced forms.
The HYB 005-01-02 immunogen is C3 found in human plasma cells.
Epitope specificity differs from that of HYB 030-08 and HAV 003-05 but slightly overlap as determined by inhibition ELISA.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.