Novus Biologicals
Manufacturer Code:NB200540AF594
Catalog # NB20540A594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 11H9 |
Immunogen | C57BL/6 thymocytes saturated with rat anti-Thy-1 monoclonal antibody of IgG2b subclass (RmT1) |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5 ARMD9 ASP C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1 complement C3 complement component 3 complement component C3 CPAMD1acylation-stimulating protein cleavage product; ASP; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3-beta-c; C3a anaphylatoxin; C3adesArg; C3bc; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component C3a; complement component C3b; epididymis secretory sperm binding protein Li 62p; prepro-C3
Gene Aliases: AHUS5; ARMD9; ASP; C3; C3a; C3b; CPAMD1; HEL-S-62p
UniProt ID: (Human) P01024
Entrez Gene ID: (Human) 718
Molecular Function: complement component cytokine defense/immunity protein enzyme modulator protease inhibitor serine protease inhibitor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.