Invitrogen
Manufacturer Code:PA521349
Catalog # PIPA521349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1448 and 1655 of (Uniprot IDNo.P01024) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5 ARMD9 ASP CPAMD1 C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1 acylation-stimulating protein cleavage product complement C3 complement component C3; ASP; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3-beta-c; C3a anaphylatoxin; C3adesArg; C3bc; Complement C3; Complement C3 alpha chain; Complement C3 beta chain; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component C3a; complement component C3b; epididymis secretory sperm binding protein Li 62p; HSE-MSF; prepro-C3
Gene Aliases: AHUS5; AI255234; ARMD9; ASP; C3; C3a; C3b; CPAMD1; HEL-S-62p; HSE-MSF; Plp
UniProt ID: (Human) P01024, (Mouse) P01027
Entrez Gene ID: (Human) 718, (Mouse) 12266
Molecular Function:
complement component
cytokine
defense/immunity protein
enzyme modulator
protease inhibitor
serine protease inhibitor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.