Novus Biologicals
Manufacturer Code:NBP29242620UL
Catalog # NB026723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-222 of human TMEM27 (NP_065716.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collectrin; collectrin kidney-specific membrane protein NX-17 transmembrane protein 27NX170610008J07Rik; kidney-specific membrane protein; Transmembrane protein 27
Gene Aliases: CLTRN; NX-17; NX17; TMEM27; UNQ679/PRO1312
UniProt ID: (Human) Q9HBJ8
Entrez Gene ID: (Human) 57393
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.