R&D Systems
Manufacturer Code:BAF4965
Catalog # BAF4965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Collectrin Glu15-Ile144 Accession # Q9ESG4 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Collectrin; collectrin kidney-specific membrane protein NX17 NX-17 TMEM27 transmembrane protein 27NX170610008J07Rik; kidney-specific membrane protein; Transmembrane protein 27
Gene Aliases: CLTRN; NX-17; NX17; TMEM27; UNQ679/PRO1312
UniProt ID: (Human) Q9HBJ8
Entrez Gene ID: (Human) 57393
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.