Novus Biologicals
Manufacturer Code:NBP267825
Catalog # NBP267825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SD83-03 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: alpha 1 (VI) chain (61 AA); Collagen 6 collagen alpha-1(VI) chain collagen VI alpha-1 polypeptide collagen type VI alpha 1 Collagen-6 human mRNA for collagen VI alpha-1 C-terminal globular domain10alpha 1 (VI) chain (61 AA) OPLL; Collagen alpha-1(VI) chain; collagen VI, alpha-1 polypeptide; collagen, type VI, alpha 1
Gene Aliases: BTHLM1; COL6A1; OPLL; UCHMD1
UniProt ID: (Human) P12109
Entrez Gene ID: (Human) 1291
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.