R&D Systems
Manufacturer Code:AF6346SP
Catalog # AF6346SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Collagen XIII Glu109-Leu668 Accession # NP_542992 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: COL13A1 collagen alpha-1(XIII) chain collagen type XIII alpha 1 FLJ42485; Collagen alpha-1(XIII) chain; collagen, type XIII, alpha 1; COLXIIIA1
Gene Aliases: CMS19; COL13A1; COLXIIIA1
UniProt ID: (Human) Q5TAT6
Entrez Gene ID: (Human) 1305
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.