Novus Biologicals
Manufacturer Code:NBP243809
Catalog # NBP243809
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6410 |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human COL11A2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Collagen alpha-2(XI) chain; collagen alpha-2(XI) chain collagen type XI alpha 2 DFNA13 DFNB53 HKE5 PARP pro-a2 chain of collagen type XI STL3; collagen, type XI, alpha 2; pro-a2 chain of collagen type XI
Gene Aliases: COL11A2; DFNA13; DFNB53; FBCG2; HKE5; PARP; STL3
UniProt ID: (Human) P13942
Entrez Gene ID: (Human) 1302
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.