Invitrogen
Manufacturer Code:MA538207
Catalog # PIMA538207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1725 |
Immunogen | A synthesized peptide derived from human Collagen VI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AI747156 alpha 1 (VI) chain (61 AA) BTHLM1 COL6A1 Col6a-1 Col6a2 Col6a-2 Collagen alpha-1(VI) chain collagen alpha-2(VI) chain collagen type VI alpha 1 collagen type VI alpha 2 collagen VI alpha-1 polypeptide collagen VI alpha-2 polypeptide collagen type VI alpha 1 collagen type VI alpha 2 human mRNA for collagen VI alpha-2 C-terminal globular domain OPLL PP3610 procollagen type VI alpha 1 procollagen type VI alpha 2 RGD1565398 UCHMD1 UCMD1; alpha 1 (VI) chain (61 AA); Collagen alpha-1(VI) chain; Collagen alpha-2(VI) chain; collagen VI, alpha-1 polypeptide; collagen VI, alpha-2 polypeptide; collagen, type VI, alpha 1; collagen, type VI, alpha 2; human mRNA for collagen VI alpha-2 C-terminal globular domain; procollagen, type VI, alpha 1; procollagen, type VI, alpha 2
Gene Aliases: AI747156; BTHLM1; Col6a-1; Col6a-2; COL6A1; COL6A2; OPLL; PP3610; RGD1565398; UCHMD1; UCMD1
UniProt ID: (Human) P12109, (Human) Q14049, (Mouse) Q04857, (Mouse) Q02788
Entrez Gene ID: (Human) 1291, (Human) 1292, (Mouse) 12833, (Rat) 294337, (Rat) 361821, (Mouse) 12834
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.