Novus Biologicals
Manufacturer Code:NBP1051180.025MG
Catalog # NB018307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E2-E4/Col5 |
Immunogen | Acid-digested pepsin soluble dog type V collagen |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha 1 type V collagen Collagen 5 collagen alpha-1(V) chain collagen type V alpha 1 Collagen-5 EC 2.7.7.6 EC 6.1.1; Collagen alpha-1(V) chain; collagen, type V, alpha 1
Gene Aliases: COL5A1; EDSC
UniProt ID: (Human) P20908
Entrez Gene ID: (Human) 1289
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.