Invitrogen
Manufacturer Code:PA523989
Catalog # PIPA523989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 54-82 amino acids from the N-terminal region of human COL5A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 1 type V collagen collagen alpha-1(V) chain; alpha1(V) collagen; Collagen alpha-1(V) chain; collagen, type V, alpha 1; pro-alpha1(V) collagen; procollagen type V alpha 1; procollagen, type V, alpha 1
Gene Aliases: AI413331; COL5A1; EDSC
UniProt ID: (Human) P20908, (Mouse) O88207
Entrez Gene ID: (Human) 1289, (Mouse) 12831
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.