Proteintech
Manufacturer Code:668241IG150UL
Catalog # 50-173-7214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H10B1 |
Immunogen | Collagen Type VI Fusion Protein Ag11809 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha 1 (VI) chain (61 AA); COL6A1 Collagen alpha 1(VI) chain Collagen Type VI collagen type VI alpha 1 OPLL; Collagen alpha-1(VI) chain; collagen VI, alpha-1 polypeptide; collagen, type VI, alpha 1
Gene Aliases: BTHLM1; COL6A1; OPLL; UCHMD1
UniProt ID: (Human) P12109
Entrez Gene ID: (Human) 1291
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.