Proteintech
Manufacturer Code:146951AP150UL
Catalog # 50-172-9648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Collagen Type I Fusion Protein Ag6281 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha 2 type I collagen COL1A2 Collagen alpha 2(I) chain Collagen Type I collagen type I alpha 2 OI4; alpha 2 type I procollagen; alpha 2(I) procollagen; alpha 2(I)-collagen; Alpha-2 type I collagen; alpha2 chain of type I collagen; Collagen alpha-2(I) chain; collagen COL1A2; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-2 chain; collagen, type I, alpha 2; osteogenesis imperfecta; procollagen, type I, alpha 2; type I procollagen
Gene Aliases: AA960264; AI325291; Col1a-2; COL1A2; Cola-2; Cola2; OI4; oim
UniProt ID: (Human) P08123, (Mouse) Q01149, (Rat) P02466
Entrez Gene ID: (Human) 1278, (Pig) 100626716, (Mouse) 12843, (Rat) 84352
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.