Invitrogen
Manufacturer Code:PA185324
Catalog # PIPA185324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A mixture of human collagen 1-5 from human placenta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AB collagen; alpha 2 type I procollagen; alpha 2(I) procollagen; alpha 2(I)-collagen; Alpha-1 type I collagen; Alpha-1 type II collagen; Alpha-2 type I collagen; alpha1 (III) collagen; alpha1(I) procollagen; Arresten; arthroophthalmopathy, progressive (Stickler syndrome); Canstatin; cartilage collagen; Chondrocalcin; COL1A1 COL1A2 COL2A1 COL3A1 COL4A1 COL4A2 COL4A3 COL4A4 COL4A5 COL4A6 COL5A1 COL5A2 COL5A3 Collagen 1 Collagen 2 Collagen 3 Collagen 4 Collagen 5 pan Collagen; COL4A1 NC1 domain; collagen alpha 1 chain type I; Collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; Collagen alpha-1(II) chain; Collagen alpha-1(III) chain; Collagen alpha-1(IV) chain; Collagen alpha-1(V) chain; Collagen alpha-2(I) chain; Collagen alpha-2(IV) chain; Collagen alpha-2(V) chain; Collagen alpha-3(IV) chain; Collagen alpha-3(V) chain; Collagen alpha-4(IV) chain; Collagen alpha-5(IV) chain; Collagen alpha-6(IV) chain; collagen I, alpha-2 polypeptide; collagen II, alpha-1 polypeptide; collagen IV, alpha-1 polypeptide; collagen IV, alpha-3 polypeptide; Collagen IV, alpha-4 polypeptide; collagen IV, alpha-5 polypeptide; collagen IV, alpha-6 polypeptide; collagen of basement membrane, alpha-1 chain; collagen of basement membrane, alpha-4 chain; collagen of basement membrane, alpha-5 chain; collagen of basement membrane, alpha-6; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen, fetal; collagen, fetal membrane, A polypeptide; collagen, type I, alpha 1; collagen, type I, alpha 2; collagen, type II, alpha 1; collagen, type III, alpha 1; collagen, type IV, alpha 1; collagen, type IV, alpha 3 (Goodpasture antigen); collagen, type IV, alpha 4; collagen, type IV, alpha 5; collagen, type IV, alpha 6; collagen, type V, alpha 1; collagen, type V, alpha 2; collagen, type V, alpha 3; dA149D17.3; dA24A23.1; dJ889N15.4 (Collagen Alpha 6(IV)); Ehlers-Danlos syndrome type IV, autosomal dominant; Goodpasture antigen; pro-(alpha)3(V) collagen; pro-alpha-1 collagen type 1; Tumstatin; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain; type V preprocollagen alpha 2 chain
Gene Aliases: ANFH; AOM; ASLN; ATS; BSVD; CA44; CA54; COL11A3; COL1A1; COL1A2; COL2A1; COL3A1; COL4A1; COL4A2; COL4A3; COL4A4; COL4A5; COL4A6; COL5A1; COL5A2; COL5A3; CXDELq22.3; DELXq22.3; DFNX6; EDS4A; EDSC; ICH; OI1; OI2; OI3; OI4; POREN2; RATOR; SEDC; STL1
UniProt ID: (Human) P02452, (Human) P08123, (Human) P02458, (Human) P02461, (Human) P02462, (Human) P08572, (Human) Q01955, (Human) P53420, (Human) P29400, (Human) Q14031, (Human) P20908, (Human) P05997, (Human) P25940
Entrez Gene ID: (Human) 1277, (Human) 1278, (Human) 1280, (Human) 1281, (Human) 1282, (Human) 1284, (Human) 1285, (Human) 1286, (Human) 1287, (Human) 1288, (Human) 1289, (Human) 1290, (Human) 50509
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.