Novus Biologicals
Manufacturer Code:NBP230450
Catalog # NBP230450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: PGKPGRPGTIQGLEGSADFLCPTNCPPGMKGPPGLQGV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 2 type IX collagen; Alpha 2 Type IX Collagen COL9A2 Collagen Alpha-2(IX) Chain Collagen IX Alpha-2 Polypeptide Collagen Type IX Alpha 2 DJ39G22.4 EDM2 MED STL5; Collagen alpha-2(IX) chain; collagen IX, alpha-2 polypeptide; collagen, type IX, alpha 2
Gene Aliases: COL9A2; DJ39G22.4; EDM2; MED; STL5
UniProt ID: (Human) Q14055
Entrez Gene ID: (Human) 1298
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.