Novus Biologicals
Manufacturer Code:NBP197716
Catalog # NBP197716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1043 |
Immunogen | 1043 is a mouse monoclonal IgG1 antibody derived by fusion of Sp 2/0 mouse myeloma cells with spleen cells from a BABL/c mouse immunized with purified human type IV collagen. The immunogen has been isolated from human placenta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha-1 chain arresten COL4A1 NC1 domain collagen alpha-1(IV) chain collagen IV alpha-1 polypeptide collagen type IV alpha 1 Collagen-4 EC 3.4.23 EC 6.3.1.2; Arresten; COL4A1 NC1 domain; Collagen alpha-1(IV) chain; collagen IV, alpha-1 polypeptide; collagen of basement membrane, alpha-1 chain; collagen, type IV, alpha 1
Gene Aliases: BSVD; COL4A1; RATOR
UniProt ID: (Human) P02462
Entrez Gene ID: (Human) 1282
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.