Invitrogen
Manufacturer Code:PA5115036
Catalog # PIPA5115036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Collagen IV alpha 2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [a]3(IV) [a]4(IV) alpha 1 type IV collagen alpha type IV collagen basement membrane alpha1(IV) collagen alpha-2 collagen type IV alpha-3 type IV collagen alpha3(IV) Arresten Bru BSVD CA44 Canstatin COL4A1 Col4a-1 COL4A1 NC1 domain Col4a2 Col4a-2 COL4A3 col4a4 Collagen 4 Collagen alpha-1(IV) chain collagen alpha-2(IV) chain Collagen alpha-3(IV) chain collagen alpha-4(IV) chain collagen IV alpha-1 polypeptide collagen IV alpha-3 polypeptide Collagen IV alpha-4 polypeptide collagen of basement membrane alpha-1 chain collagen of basement membrane alpha-4 chain collagen type IV alpha 1 chain collagen type IV alpha 2 collagen type IV alpha 3 chain collagen type IV alpha 4 chain collagen type IV alpha3 chain collagen type IV alpha 1 collagen type IV alpha 2 collagen type IV alpha 3 collagen type IV alpha 3 (Goodpasture antigen) collagen type IV alpha 4 Del(8)44H Del(8)Bru44H E130010M05Rik Goodpasture antigen ICH POREN2 procollagen alpha1(IV) procollagen type IV alpha 1 procollagen type IV alpha 2 procollagen type IV alpha 3 procollagen type IV alpha 4 RATOR Raw retinal anterior wiring Svc Tumstatin Type IV Collagen; alpha 1 type IV collagen; alpha-2 collagen type IV; alpha1(IV) collagen; Arresten; Canstatin; COL4A1 NC1 domain; col4a3; col4a4; Collagen alpha-1(IV) chain; Collagen alpha-2(IV) chain; Collagen alpha-3(IV) chain; Collagen alpha-4(IV) chain; collagen IV, alpha-1 polypeptide; collagen IV, alpha-3 polypeptide; Collagen IV, alpha-4 polypeptide; collagen of basement membrane, alpha-1 chain; collagen of basement membrane, alpha-4 chain; collagen type IV alpha3 chain; collagen, type IV, alpha 1; collagen, type IV, alpha 3 (Goodpasture antigen); collagen, type IV, alpha 4; Del(8)Bru44H; Goodpasture antigen; procollagen, type IV, alpha 1; procollagen, type IV, alpha 2; procollagen, type IV, alpha 3; procollagen, type IV, alpha 4; retinal anterior wiring; Tumstatin
Gene Aliases: [a]3(IV); [a]4(IV); alpha3(IV); Bru; BSVD; CA44; Col4a-1; Col4a-2; COL4A1; COL4A2; COL4A3; COL4A4; Del(8)44H; E130010M05Rik; ICH; POREN2; RATOR; Raw; Svc
UniProt ID: (Human) P02462, (Human) P08572, (Human) Q01955, (Human) P53420, (Mouse) P02463, (Mouse) P08122, (Mouse) Q9QZS0, (Mouse) Q9QZR9
Entrez Gene ID: (Human) 1282, (Human) 1284, (Human) 1285, (Human) 1286, (Rat) 290905, (Mouse) 12826, (Rat) 306628, (Mouse) 12827, (Rat) 363265, (Mouse) 12828, (Mouse) 12829, (Rat) 301562
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.