Novus Biologicals
Manufacturer Code:NBP105119F
Catalog # NBP105119F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E7-D7/Col3 |
Immunogen | Acid-digested pepsin soluble human type III collagen |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha1 (III) collagen; alpha1 (III) collagen Collagen 3 collagen alpha-1(III) chain collagen fetal collagen type III alpha 1 Collagen-3 EDS4A Ehlers-Danlos syndrome type IV autosomal dominant FLJ34534; Collagen alpha-1(III) chain; collagen, fetal; collagen, type III, alpha 1; Ehlers-Danlos syndrome type IV, autosomal dominant
Gene Aliases: COL3A1; EDS4A
UniProt ID: (Human) P02461
Entrez Gene ID: (Human) 1281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.